missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, TRC

Product Code. 17994185
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994185 25 mg 25mg
30008001 50 mg 50mg
17992615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994185 Supplier Toronto Research Chemicals Supplier No. TRCA72650025MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-36-5
Molecular Formula C15 H10 O5
Molecular Weight (g/mol) 270.24
Synonym Flavone, 4',5,7-trihydroxy- (8CI), 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',5,7-Trihydroxyflavone, 5,7,4'-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one, 5,7-Dihydroxy-2-p-hydroxyphenyl-4-chromenone, Apegenin, Apigenin, Apigenine, Apigenol, C.I. Natural Yellow 1, Chamomile, LY 080400, NSC 83244, Pelargidenon 1449, ST 056301, UCCF 031, Versulin, 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-(4-hydroxyphenyl)-
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H
Formula Weight 270.0528
Percent Purity >95%
Chemical Name or Material Apigenin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.