missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Apilimod dimesylate
GREENER_CHOICE

Product Code. 18767466
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767466 25 mg 25mg
18799956 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18767466 Supplier Tocris Bioscience™ Supplier No. 7283/25
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective PIKfyve inhibitor

Apilimod dimesylate is a potent and selective PIKfyve inhibitor (IC50 = 14 nM). Exhibits no significant activity at other lipid kinases and protein kinases, including PIP4K, PIP5K, mTOR, PI3K and PI4K. Inhibits cellular entry by SARS-CoV-2, MERS-CoV and MHV S pseudovirions. Reduces number of SARS-CoV-2-infected hiPSC-derived pneumocyte-like cells by 85% and inhibits SARS-CoV-2 replication in donor lung tissue. Also exhibits selective cytotoxicity in B-cell non-Hodgkin lymphoma compared with normal cells. Inhibits production of IL-12 and IL-23. Anti-inflammatory. Water-soluble.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(6-(2-(3-Methylbenzylidene)hydrazinyl)-2-(2-(pyridin-2-yl)ethoxy)pyrimidin-4-yl)morpholine dimethanesulfonate
CAS 870087-36-8
Quantity 25 mg
Target PIKFyve/PIP5K3
Molecular Formula C23H26N6O20.2 CH3SO3H
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.