missing translation for 'onlineSavingsMsg'
Learn More

Apilimod mesylate, MedChemExpress

Product Code. 16476118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476118 10 mg 10mg
16468717 5 mg 5mg
16462257 50 mg 50mg
16416516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476118 Supplier Medchem Express Supplier No. HY14644A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Apilimod (STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Apilimod is a potent and highly selective PIKfyve inhibitor.

Apilimod (STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC50s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Apilimod is a potent and highly selective PIKfyve inhibitor.

Specifications

Chemical Name or Material Apilimod mesylate
CAS 870087-36-8
Color White
Purity Grade Notes Research
Molecular Formula C25H34N6O8S2
Quantity 10 mg
Synonym STA 5326 mesylate
Solubility Information H2O : 100 mg/mL (163.75 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (20.47 mM; Need ultrasonic)
SMILES CC1=CC=CC(/C=N/NC2=CC(N3CCOCC3)=NC(OCCC4=CC=CC=N4)=N2)=C1.CS(=O)(O)=O.CS(=O)(O)=O
Molecular Weight (g/mol) 610.7
Formula Weight 610.7
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.