missing translation for 'onlineSavingsMsg'
Learn More

BD Biosciences™ APO-BRDU™ Kit (RUO)

Product Code. 15828688 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
60 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15828688 60 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15828688 Supplier BD Biosciences™ Supplier No. 556405
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry.

  • Two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry.
  • Sufficient reagents are provided to process 60 cell suspensions.
  • The kit includes 5 mL of positive and 5 mL of negative control cell suspensions, approximately 1 x 10<sup>6</sup> cells per mL in 70% (v/v) ethanol.
  • The control cells are derived from a human lymphoma cell line and have been fixed as described in the Fixation Protocol.
TRUSTED_SUSTAINABILITY

Specifications

Description 60 Tests
Quantity 60 Tests
Storage Buffer 0.05% sodium azide
Content And Storage One kit/two parts: Part A - 0.30 mL bottle of FITC-Labeled Anti-BrdU mAb, 30 mL bottle of PI/RNase Staining Buffer, 0.6 mL vial of Reaction Buffer, 126 mL bottle of Rinsing Buffer, 120 mL bottle of Wash Buffer. Part B - 0.48 mL bottle of Br-dUTP, 5 mL bottle of Negative Control Cells, 5 mL bottle of Positive Control Cells, 0.045 mL bottle of TdT Enzyme.. Part A (Comp. No. 51-6576AK) is shipped at ambient temperature and should be stored at 4°C upon arrival. Part B (Comp. No. 51-6576BK) is shipped at -80°C on dry ice, and should be stored at -20°C upon arrival.
For Use With (Application) Labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry
compliance-icons compliance-icons compliance-icons
Product Identifier
  • APO-BRDU Kit
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, cacodylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.