missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ APO-BrdU™ TUNEL Assay Kit, with Alexa Fluor™ 488 Anti-BrdU

Product Code. 10043102
Change view
Click to view available options
Quantity:
60 Assays
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043102 60 Assays 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10043102 Supplier Molecular Probes™ Supplier No. A23210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

APO-BrdU™ TUNEL Assay Kit, with Alexa Fluor™ 488 Anti-BrdU

In collaboration with Phoenix Flow Systems, we offer the APO™-BrdU TUNEL Assay Kit which provides all the materials necessary to label and detect the DNA strand breaks of apoptotic cells. When DNA strands are cleaved or nicked by nucleases, a large number of 3´hydroxyl ends are exposed. In the APO-BrdU assay, these ends are labeled with BrdUTP and terminal deoxynucleotidyl transferase (TdT) using the TUNEL technique. Once incorporated ino the DNA, BrdU is detected using a bright and photostable green-fluorescent Alexa Fluor™ 488 dye-labeled anti-BrdU antibody.
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Contains positive control cells (5 mL) and negative control cells (5 mL) of a fixed human lymphoma cell line, 1 vial of TdT (45 μL), 1 vial of BrdUTP (480 μL), 1 vial of anti-BrdU Ms mAb Alexa Fluor™ 488 conjugate (350 μL), 1 bottle of PI/Rnase A staining buffer (30 mL), 1 vial of reaction buffer (0.6 mL), 1 bottle of wash buffer (120 mL) and 1 bottle of rinse buffer (120 mL).

Store as directed.
Detection Method Fluorescence
Format Tube
Excitation/Emission 490/525
Flow Cytometer Laser Lines 488
For Use With (Application) Flow Cytometry
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Label or Dye Alexa Fluor™ 488
Product Type TUNEL Assay Kit
Product Line APO-BrdU
Quantity 60 Assays
Species Mouse
Target BrdU
Label Type Alexa Fluor Dyes
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • APO-BrdU TUNEL Assay Kit, with Alexa Fluor 488 Anti-BrdU
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.