missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Apocynin

Product Code. 30001626
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001626 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001626 Supplier R&D Systems™ Supplier No. 4663/50
  • ONLINE EXCLUSIVE PRICE
    91.05 €
    81.90 € / 50mg
    Save 9.15 €
    10% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. NADPH-oxidase inhibitor

Apocynin is an NADPH oxidase inhibitor. Demonstrates anti-inflammatory properties in a mouse model of carrageenan-induced pleurisy. Apocynin is bioactivated by intracellular peroxidases; acts as an antioxidant in endothelial cells and vascular smooth muscle cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 498-02-2
Quantity 50 mg
Product Line Tocris
Molecular Formula C9H10O3
Formula Weight 166.17
Purity >99%
Molecular Weight (g/mol) 166.18
Chemical Name or Material Apocynin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.