missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ApoDETECT Annexin V-FITC Kit

Product Code. 10565883
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10565883 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10565883 Supplier Invitrogen™ Supplier No. 331200
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ApoDETECT Annexin V-FITC Kit

The ApoDETECT Annexin V-FITC Kit provides all the reagents needed to detect the apoptotic cells. Annexin V is a Ca2+-dependent phospholipid binding protein. This protein can bind to a variety of phospholipids, but it has the highest affinity for phosphatidylserine (PS). The ApoDETECT Annexin V-FITC Kit binds to negatively charged phospholipid surfaces in a Ca2+-dependent manner. It also prevents the formation of the prothrombinase complex, thereby inhibiting formation of thrombin (anticoagulant activity). The ApoDETECT Annexin V-FITC Kit can be used to detect apoptotic cells by flow cytometry or immunofluorescent cytology.

Kit Attributes:

  • Applications: Validated application for ApoDETECT Annexin V-FITC Kit is detecting apoptotic cells by flow cytometry or immunofluorescent cytology.
  • Format: Vial(s)
  • Detection Method: Fluorescence microscopy
  • Product Size: One kit

    In normal cells phosphatidylserine (PS) is located on the inner leaflet of the plasma membrane. During the early stages of apoptosis, PS is translocated to the outer layer and is exposed on the external surface of the cell. This early event in apoptosis can be detected by using a sensitive method in which to detect PS exposure. Translocation of PS to the external surface of the plasma membrane is not a unique property of apoptotic cells, as this phenomenon also occurs during cell necrosis. However, during apoptosis, the cell membrane remains intact; whereas, during necrosis, the cell becomes leaky and loses its integrity. Therefore, it is necessary to assess membrane integrity together with PS translocation.

    Annexin V is a Ca2+-dependent phospholipid binding protein. This protein can bind to a variety of phospholipids, but it has the highest affinity for phosphatidylserine (PS). Based on its affinity for PS, Annexin V can be utilized as a sensitive probe for cell surface exposure of PS. To use the Annexin V protein as a probe for apoptotic cells, the protein has been labeled with fluorescein isothiocyanate (FITC). In this form, the protein can be used directly for quantification of apoptotic cells. The measurement of Annexin V binding when performed simultaneously with a dye exclusion test (such as propidium iodide) can be used to effectively discriminate between apoptotic and necrotic cells.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Quantity 1 kit
    Format Vial
    Product Type Annexin V-FITC Kit
    Content And Storage Store in refrigerator (2–8°C).
    Shipping Condition Wet Ice
    For Use With (Application) Flow Cytometry
    Conjugate FITC
    For Use With (Equipment) Fluorescence Microscope, Flow Cytometer

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.