missing translation for 'onlineSavingsMsg'
Learn More

Apovincaminic Acid-d4, TRC

Product Code. 30331849
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331849 10 mg 10mg
30332290 1 mg 1mg
30347939 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30331849 Supplier Toronto Research Chemicals Supplier No. TRCA72925210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Apovincaminic Acid-d4
Molecular Formula C20H18D4N2O2
Quantity 10 mg
Synonym (13aS,13bS)-13a-Ethyl-2,3,5,6,13a,13b-hexahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylic Acid-d4, Apovincamin-22-oic Acid-d4, (3α,16α)-Eburnamenine-14-carboxylic Acid-d4, (+)-Apovincaminic Acid-d4, (+)-cis-Apovincaminic Acid-d4
InChI Formula InChI=1S/C20H22N2O2/c1-2-20-9-5-10-21-11-8-14-13-6-3-4-7-15(13)22(17(14)18(20)21)16(12-20)19(23)24/h3-4,6-7,12,18H,2,5,8-11H2,1H3,(H,23,24)/t18-,20+/m1/s1/i3D,4D,6D,7D
SMILES [H][C@@]1([C@]2(CC)C=C3C(O)=O)N(C([2H])([H])C([2H])([H])C2)CCC4=C1N3C5=C([2H])C=C([2H])C=C54
IUPAC Name (15S,19S)-3,4,5,6-tetradeuterio-15-ethyl-1,11-diazapentacyclo[9.6.2.02,7.08,18.015,19]nonadeca-2,4,6,8(18),16-pentaene-17-carboxylic acid
Molecular Weight (g/mol) 326.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.