missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq™ 360 Buffer, 25 mM MgCl2 and 360 GC Enhancer
GREENER_CHOICE

Product Code. 10489014 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10489014 1.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10489014 Supplier Applied Biosystems™ Supplier No. 4398848
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

AmpliTaq 360 Buffer kit is supplied 1.5mL of each 10X AmpliTaq™ 360 Buffer, 25mM M2Cl2, and 360 GC Enhancer. Typically, 1.5mL of each of AmpliTaq 360 Buffer, 25mM MgCl2, and 360 GC Enhancer is sufficient for use with 250U of enzyme.. All solutions are prepared in deionized, filter-sterilized water.

The Applied Biosystems AmpliTaq 360 Buffer Kit is used in conjection with the AmpliTaq 360 DNA polymerase for those few experiments when the reagents supplied with the enzyme are not sufficient for the researcher's needs.

The Applied Biosystems AmpliTaq 360 Buffer Kit is used in conjection with the AmpliTaq 360 DNA Polymerase for situations when the reagents supplied with the enzyme are not sufficient for the experimental needs. When used with or without the optional 360 GC Enhancer, this buffer amplifies a vast range of DNA sequence contexts. AmpliTaq 360 DNA Polymerase delivers 360° coverage for a full range of targets.

The AmpliTaq 360 Buffer Kit is designed for use with challenging targets including AT-rich, GC-rich, primer-dimer forming amplicons, homopolymer repeats, and amplicons that pose sequencing challenges. Amplicons that previously required specialized enzymes and reaction conditions can be reproducibly amplified with a single reagent under standardized conditions. Competitive benchmarking across more than 40 amplicons distinguishes AmpliTaq Gold 360 Polymerase as a superior-performing enzyme, ensuring the highest probability of success for the amplification of both everyday and challenging targets.

Features

  • Optimized for the broad range of targets
  • Robust amplification of GC-rich sequences with 360 GC Enhancer
  • Achieves high quality sequencing data

Notes

  • GC-rich regions are especially poorly amplified with competitor DNA polymerases and the original AmpliTaq Gold, while AmpliTaq Gold 360 provides successful, robust amplification.
  • See user's manual or package insert for limited label license and trademark information.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • 10X AmpliTaq 360 Buffer, 1.5 mL
• 25 mM MgCl2, 1.5 mL
• 360 GC Enhancer, 1.5 mL

Store at -15°C to -25°C.
Product is guaranteed up to 12 months as indicated by the expiration date.
GC-Rich PCR Performance High
Polymerase AmpliTaq 360 DNA Polymerase
Reaction Speed Standard
Product Type GC Enhancer
Quantity 1.5 mL
Shipping Condition Room Temp or Wet Ice
For Use With (Application) Standard PCR
Reaction Format Kit
Volume 1.5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.