missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 Buffer Kit
GREENER_CHOICE

Product Code. 10504055 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10504055 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10504055 Supplier Applied Biosystems™ Supplier No. 4398853
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

The AmpliTaq Gold 360 Buffer Kit includes 1.5mL (each) of 10X AmpliTaq Gold 360 Buffer, 25mM MgCl2, and 360 GC Enhancer. All solutions are prepared in deionized, filter-sterilized water.

The AmpliTaq Gold 360 Buffer Kit is used in conjunction with AmpliTaq Gold 360 DNA Polymerase and is intended for those occasions when the quantity of reagents supplied with the enzyme are not sufficient.

The AmpliTaq Gold 360 Buffer Kit is used in conjunction with AmpliTaq Gold 360 DNA Polymerase and is intended for use on occasions when the quantity of reagents supplied with the enzyme are not sufficient.

This kit includes 1.5 mL (each) of:

  • 10X AmpliTaq Gold 360 Buffer
  • 25 mM MgCl2
  • 360 GC Enhancer

Typically, 1.5 mL is sufficient for use with 250 U of enzyme.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage • 10X AmpliTaq Gold 360 Buffer, 1 x 1.5 mL
• 25 mM MgCl2, 1 x 1.5 mL
• 360 GC Enhancer, 1 x 1.5 mL

Store at -15° to -25°C.
Format Kit
Polymerase AmpliTaq Gold 360 DNA Polymerase
Product Type PCR Buffer
Quantity 1 Kit
Shipping Condition Room Temp or Wet Ice
For Use With (Application) Cloning
Fidelity (vs. Taq) 1X
Volume 1.5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.