missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase, LD (Low DNA) with Gold Buffer and MgCl2

Product Code. 10258364 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1000 Units
250 Units
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10258364 1000 Units 1 set
10055114 250 Units 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10258364 Supplier Applied Biosystems™ Supplier No. 4338857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

Contains AmpliTaq Gold DNA Polymerase, LD (250 units), GeneAmp 10X PCR Gold Buffer, and MgCl2 Solution.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA) is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

Applied Biosystems AmpliTaq Gold DNA Polymerase, LD (Low DNA), is a highly purified version of AmpliTaq Gold DNA Polymerase that significantly reduces contaminating bacterial DNA sequences commonly present in recombinant protein preparations.

This highly purified enzyme preparation gives you more confidence in your results and makes advanced PCR techniques quick and easy. When combined with the supplied GeneAmp 10X PCR Gold Buffer, AmpliTaq Gold DNA Polymerase, LD, provides a flexible hot-start system that is suitable for all PCR applications that require the elimination of bacterial DNA contamination.

Features

  • Highly purified enzyme with lower levels of bacterial DNA
  • Minimized non-target, false-positive PCR products when amplifying bacterial sequences
  • Chemically modified hot-start technique eliminates biological contamination
  • 10X PCR Gold Buffer promotes fast enzyme activation and robust PCR amplification
  • Enhanced specificity with time-release PCR for low-copy target amplification
  • Improved yield and success rate for target amplification

Notes

  • AmpliTaq Gold DNA Polymerase, LD, is recommended for PCR applications that require low background levels of bacterial DNA.
  • See user's manual or package insert for limited label license and trademark information.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq Gold DNA Polymerase, LD (5 U/μL), 1 x 200 μL
• 10X PCR Gold Buffer, 4 x 1.5 mL
• 25 mM MgCl2, 4 x 1.5 mL

Store at -15°C to -30°C.
Format Tube
GC-Rich PCR Performance Low
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Purity or Quality Grade LD (Low DNA)
Quantity 1000 Units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 800 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.