missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer II & MgCl2

Product Code. 10219164 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 x 1.5 mL
6 x 1.5 mL
2 x 75 mL
Unit Size:
150mL
Each
This item is not returnable. View return policy

Product Code. 10219164

Brand: Applied Biosystems™ N8080130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

2 x 75mL each of GeneAmp™ 10X PCR Buffer II and MgCl2 sufficient for 100 PCR amplifications of 100μL each.

Applied Biosystems GeneAmp 10X PCR Buffer II is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Applied Biosystems™ GeneAmp™ 10X PCR Buffer II is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification. The provided MgCl2 solution offers versatility for optimizing PCR.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes
  • Separate MgCl2 solution offers versatility

GeneAmp™ 10X PCR Buffer II, when diluted 1:10, provides the optimal pH and ionic strength for PCR amplification. GeneAmp™ 10X PCR Buffer II is magnesium ion free. A separate MgCl2 solution is provided. When used together, GeneAmp™ 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage • GeneAmp 10X PCR Buffer II, 6 x 1.5 mL
• MgCl2, 6 x 1.5 mL

Store at -15°C to -25°C.
Polymerase AmpliTaq DNA Polymerase
Product Type PCR Buffer
Quantity 6 x 1.5 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
No. of Reactions 100 Reactions
Reaction Format Kit
Volume 9 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis