missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer II & MgCl2

Product Code. 10229724 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 1.5 mL
6 x 1.5 mL
2 x 75 mL
Unit Size:
150mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10229724 2 x 75 mL 150mL
10219774 1 x 1.5 mL Each
10219164 6 x 1.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10229724 Supplier Applied Biosystems™ Supplier No. 4379878
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

2 x 75mL each of GeneAmp™ 10X PCR Buffer II and MgCl2 sufficient for 100 PCR amplifications of 100μL each.

Applied Biosystems GeneAmp 10X PCR Buffer II is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Applied Biosystems™ GeneAmp™ 10X PCR Buffer II is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification. The provided MgCl2 solution offers versatility for optimizing PCR.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes
  • Separate MgCl2 solution offers versatility

GeneAmp™ 10X PCR Buffer II, when diluted 1:10, provides the optimal pH and ionic strength for PCR amplification. GeneAmp™ 10X PCR Buffer II is magnesium ion free. A separate MgCl2 solution is provided. When used together, GeneAmp™ 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Content And Storage • GeneAmp 10X PCR Buffer II, 2 x 75 mL
• MgCl2, 2 x 75 mL

Store at -15°C to -25°C.
Polymerase AmpliTaq DNA Polymerase
Product Type PCR Buffer
Quantity 2 x 75 mL
Shipping Condition Wet Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Reaction Format Kit
Volume 150 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.