missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM)

Product Code. 10085714
Change view
Click to view available options
Quantity:
1 mL
12 x 1 mL
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10085714 1 mL Each
10269624 12 x 1 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10085714 Supplier Applied Biosystems™ Supplier No. N8080261
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 μL each. Each kit contains 12 x 1 mL vials.

Key advantages of GeneAmp dNTPs

  • High purity dNTPs are ideal for use in PCR reactions or other applications
  • Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
  • Premixed dNTP blends are available in various concentrations, offering ease of use

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
Content And Storage GeneAmp dNTP Blend, 100 mM (1 mL)

Store in a constant temperature freezer at –20°C.
Form Liquid
Product Line GeneAmp
Quantity 1 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.