missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM)

Product Code. p-4925099 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 mL
12 x 1 mL
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy

Product Code. 10085714

Brand: Applied Biosystems™ N8080261

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 µL each. Each kit contains 12 x 1 mL vials.

Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 mM
Content And Storage GeneAmp dNTP Blend, 100 mM (1 mL)

Store in a constant temperature freezer at –20°C.
Form Liquid
Product Line GeneAmp
Quantity 1 mL
Shipping Condition Dry Ice
For Use With (Application) Standard PCR

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.