missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ Fast PCR Master Mix (2X), with or without protocol

Product Code. 10526745 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
250 Reactions (with protocol)
250 Reactions (withithout protocol)
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10526745 250 Reactions (withithout protocol) Each
10229534 250 Reactions (with protocol) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10526745 Supplier Applied Biosystems™ Supplier No. 4359187
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

The GeneAmp Fast PCR Master Mix (2X) contains two tubes (1.25mL each) sufficient for 125 20μL PCR reactions. The Fast PCR Master Mix can also be ordered with a protocol.

The GeneAmp Fast PCR Master Mix, with its sensitive, pre-mixed hot-start chemistries, reduces PCR reaction time to as little as 25 minutes when used with the Veriti 96-Well Thermal Cycler.

Applied Biosystems GeneAmp Fast PCR Master Mix reduces PCR reaction time to as little as 25 minutes when used with the Veriti 96-well thermal cycler. The sensitive, premixed hot-start chemistries of this master mix enable quick experiment setup and by generating greater than 90% 'A' overhang, the cloning process is further simplified.

GeneAmp Fast PCR Master Mix may be used to detect a single copy of a gene in 10 ng of human genomic DNA or as little as 10 copies of an HIV target.

Features

  • Quick reaction setup with premixed reagents
  • Gene detection with as little as 10 ng of human genomic DNA
  • Reproducible, specific, and sensitive results

Notes

  • The Applied Biosystems Fast PCR System, consisting of the premixed GeneAmp Fast PCR Master Mix, specially designed microplates and tubes, and the Veriti 96-well thermal cycler (0.1 mL or 0.2 mL), considerably reduces PCR reaction times.
  • GeneAmp Fast PCR Master Mix is provided at a convienient 2X concentration with only the addition of DNA template and primers required.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage • 2X GeneAmp Fast PCR Master Mix, 2 x 1.25 mL tubes

Store at 2°C to 8°C.
Format Tube
GC-Rich PCR Performance Medium
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast
Exonuclease Activity 5' - 3'
Product Type Fast PCR Master Mix
Quantity 250 Reactions (withithout protocol)
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 250 Reactions
Overhang 3'-A
Reaction Format SuperMix or Master Mix
Size (Final Product) 2 kb or less
Starting Material DNA
Volume 0.1 mL or 0.2 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.