missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700

Product Code. 15860383 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 pad
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15860383 1 pad Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15860383 Supplier Applied Biosystems™ Supplier No. 4426825
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Properly and evenly positions up to eight TaqMan™ OpenArray™ Genotyping Plates for PCR amplification

It is intended for use with the Dual Flat Block GeneAmp™ PCR System 9700 System and OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700.

  • Platform: GeneAmp™ 9700
  • Product Size: 1 pad
  • Accessory Type: Compression Pad
  • Plate Compatibility: OpenArray™ Plates
  • Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Required compression pad for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system
For Use With (Equipment) GeneAmp 9700
Product Line GeneAmp, OpenArray
Product Type Plate Compression Pad
For Use With (Application) PCR
Shipping Condition Room Temperature
Quantity 1 pad
Plate Compatibility OpenArray™ Plates

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.