missing translation for 'onlineSavingsMsg'
Learn More

MeltDoctor™ MeltDoctor™ HRM Master Mix

Product Code. 10351235 Shop All Applied Biosystems Products
Click to view available options
Quantity:
5 mL
50 mL
5 x 5 mL
10 x 5 mL
Unit Size:
25mL
50mL
Each
This item is not returnable. View return policy

Product Code. 10351235

Brand: MeltDoctor™ 4415440

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

Provided in a convenient 2X mix, containing: AmpliTaq Gold™ 360 DNA Polymerase, MeltDoctor HRM Dye, a dNTP blend including dUTP, Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results

Requires

Template and primers

Contains all components (excluding template and primers) formulated for superior HRM performance across a wide range of genomic targets.

High Resolution Melting (HRM) analysis is a post-PCR analysis method used to identify genetic variation in nucleic acid sequences. Unlike some competitive mixes, the MeltDoctor™ HRM Master Mix does not require additional mixing prior to use, and was developed and optimized solely for HRM applications.

  • Low background fluorescence
  • High brightness in the presence of double-stranded DNA
  • Minimal temperature shift of DNA melting due to dye binding
  • Thermal stability to tolerate PCR cycling conditions
  • No inhibition of polymerase activity, resulting in high PCR efficiency
  • A dNTP blend including dUTP, which minimizes carryover contamination by allowing amplicon degradation by uracil DNA glycosylase (UDG) in subsequent PCR reactions

Perform Mutation Scanning:
  • HRM analysis can be used to scan for mutations in target genes for the identification of variant samples prior to sequencing analysis
  • As a mutation scanning technique, HRM offers significant advantages over conventional methods such as Denaturing High- Performance Liquid Chromatography (DHPLC) and Denaturing Gradient Gel Electrophoresis (DGGE)

Advantages of HRM for mutation scanning
  • Low reagent consumption, with little waste: HRM requires only a 20μL PCR reaction for analysis of each sample, eliminating the need for HPLC solvents or DGGE gels
  • Simple, fast workflow: no additional instrumentation is required after PCR amplification;A high-resolution melt step can be simply added to the end of the PCR profile for immediate analysis
  • Fast optimization: unlike DHPLC, thermal optimization is not required
  • Low sample consumption: following HRM analysis, the PCR product can be used directly in a Sanger sequencing reaction
    Quality Components
    • Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results
    • AmpliTaq Gold™ 360 DNA Polymerase, a highly purified DNA polymerase that provides hot-start performance, minimizing non-specific product formation and enabling reactions to be set up at room temperature
    • MeltDoctor™ HRM Dye, a stabilized form of the fluorescent SYTO™ 9 double-stranded nucleic acid stain developed by Molecular Probes

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Components of the MeltDoctor™ HRM Master Mix, provided in a convenient 2X mix, include: AmpliTaq Gold™ 360 DNA Polymerase, MeltDoctor*trade; HRM Dye, a dNTP blend including dUTP, Magnesium salts and other buffer components, precisely formulated to obtain optimal HRM results.

Store at -15 °C to -25 °C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method HRM
Form Liquid
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Gold 360 DNA Polymerase
Reaction Speed Fast
Technique HRM (High Resolution Melting) Analysis, Mutation Scanning
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
Genotyping Target SNPs (Unknown or Numerous), Epigenetic Targets
Label or Dye High Resolution Melting (HRM), SYTO 9
Product Line MeltDoctor™
Product Type HRM Master Mix
Quantity 5 mL
Shipping Condition Wet Ice
For Use With (Application) Genotyping
No. of Reactions 500 Reactions
Sample Type DNA (Genomic), cDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.