missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ 8-Cap Strip, assorted colors

Product Code. 10713625 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
300 strips
Unit Size:
Pack of 300
Product Code. Quantity unitSize
10713625 300 strips Pack of 300
1 options
This item is not returnable. View return policy

Product Code. 10713625

Brand: Applied Biosystems™ N8010835

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Designed to fit on MicroAmp reaction tubes, tube strips, and 96-well plates

MicroAmp™ Cap Strips have a domed profile and are suitable for end-point PCR. 8-cap strips available in clear or an assorted set of red, orange, blue, and green.

  • Will not interfere with sample reading
  • Provides tight seal to minimize evaporation
  • Easy to apply and remove
Minimizes Evaporation
  • Use MicroAmp Caps Strips to seal tubes and reduce the possibility of contamination in sample wells and help ensure reliable results
  • Easy to Use
    • Use MicroAmp Cap Installing Tool (sold separately) to push caps down and form tight seal
    • When ready, just peel away caps using tabs at end
    Colored for Easy Organization
    • Available in different colors to help track tubes during assay preparation
    • For real-time PCR applications,use MicroAmp Optical 8-Cap Strips (Mfr. No. 4323032) with flat-top profile

    PCR, PCR & Real-Time PCR

    TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance Certified DNA/Rnase Free
    Color Assorted
    Volume (Metric) 0.2 mL
    Quantity 300 strips
    For Use With (Equipment) Thermal Cyclers
    Product Line MicroAmp™ 
    Material Polypropylene
    Content And Storage Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates. Assorted colors.
    Shipping Condition Room Temperature
    Closure Type Domed Cap
    No. of Wells 96
    Format 200 Strip Caps
    Product Type 8-Strip Cap
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.