missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Fast SYBR™ Green Master Mix

Product Code. 10631376 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
Unit Size:
1 set
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10631376 10 x 5 mL Pack of 10
10381275 1 x 1 mL Each
10459604 1 x 5 mL Each
10556555 2 x 5 mL 1 set
10566555 5 x 5 mL 1 set
10447494 1 x 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10631376 Supplier Applied Biosystems™ Supplier No. 4385618

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SYBR Green I Dye, AmpliTaq Gold Fast DNA Polymerase LD, dNTPs with dUTP/dTTP blend, Passive Reference I and optimized buffer components

Requires

Template and primers

Designed for speed, SYBR™ Green Master Mix provides a fast, reliable, and cost-effective solution for RT-PCR application without compromising sensitivity, specificity, dynamic range, or PCR efficiency.

  • A fast, reliable, and cost-effective solution for real-time PCR applications without compromising sensitivity, specificity, dynamic range, or PCR efficiency
  • Real-time PCR results in as fast as 35 minutes
  • Capable of detecting very low copies of target
  • Minimizes primer-dimer and non-specific amplification
  • Consistent amplification across a wide dynamic range
  • Contains all of the components, excluding the template and primers, in a convenient 2X master mix
  • Store at -5°C to -30°C

Includes the following components in an optimized buffer:

  • AmpliTaq™ Fast DNA Polymerase, UP, a highly purified DNA polymerase designed to allow instant hot start
    • Minimizes non-specific product formation and enables reactions to be set up at room temperature
  • SYBR™ Green I dye to enable detection of double-stranded DNA
  • Deoxynucleotides (dNTPs) to help maintain optimal PCR results
  • Uracil-DNA Glycosylase (UDG) designed to reduce carryover contamination
  • Passive internal reference based on proprietary ROX™ dye to enable increased precision

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage 10 × 5 mL vials. The 2X mix contains SYBR™ Green 1 Dye, AmpliTaq Gold™ Fast DNA Polymerase LD, dNTPs with dUTP⁄dTTP blend, Passive Reference 1, and optimized buffer components. Sufficient for 5000 reactions of 20 μL each.

Store at -5 to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Detection Method SYBR
Format Vial
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq Fast DNA Polymerase, AmpliTaq Fast UP
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Label or Dye SYBR Green
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR
Product Type Real Time PCR SYBR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 10 x 5 mL
Sufficient For 5000 Reactions of 20 μL Each
For Use With (Application) Gene Expression
No. of Reactions 5000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 10 x 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.