missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SYBR™ Select Master Mix

Product Code. 13276519 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 5 mL
5 x 5 mL
10 x 5 mL
1 x 50 mL
Unit Size:
100 reactions
1000 reactions
500 reactions
5000 reactions
Pack of 10
Pack of 2500
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13276519 2 x 5 mL 1000 reactions
13246519 1 x 1 mL 100 reactions
13256519 1 x 5 mL 500 reactions
13266519 5 x 5 mL Pack of 2500
13236529 10 x 5 mL Pack of 10
13286519 1 x 50 mL 5000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 13276519 Supplier Applied Biosystems™ Supplier No. 4472918

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Includes

2X mix contains SYBR GreenER dye, AmpliTaq DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components.

Cost-effective solution for real-time PCR applications.

  • Specific - minimize primer-dimer and non-specific amplification
  • Reproducible and sensitive - consistent amplification across a wide dynamic range
  • Bright - contains SYBR™ GreenER™ dye for maximum brightness
  • Carry-over contamination control - contains heat-labile UDG
  • Can be used in either standard or fast cycling mode

Formulated for Maximum Specificity:
  • SYBR™ Select Master Mix contains all the components needed for your real-time PCR reaction, except the template and primers, in a convenient 2X concentration premix
  • The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity

Obtain Reproducible Results Across a Wide Dynamic Range:
  • SYBR™ Select Master Mix is specially formulated to provide robust results from 100ng to 0.1pg cDNA per reaction and can reliably detect a single copy of genomic DNA

Contains SYBR™ GreenER™ Dye and Heat-labile UDG:
  • SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye resulting in brighter signals
  • Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control

Instrument Compatibility:
  • SYBR™ Select Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments except the 7900HT real-time PCR system
  • It is also compatible with the Bio-Rad IQ™5, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems

Order Info

Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems.
Detection Method SYBR
Form Liquid
Format Vial
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Fast Mode, StepOne™, Standard Mode, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System, AB StepOnePlus™, Fast Mode, AB StepOne™, Standard Mode, AB 7500, Fast Mode, AB 7500, Standard Mode, AB StepOne™, Fast Mode, AB StepOnePlus™, Standard Mode
Label or Dye SYBR GreenER
Passive Reference Dye ROX (Pre-mixed)
Product Line SYBR
Product Type Real Time PCR SYBR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 2 x 5 mL
Sufficient For 100 Reactions of 20 μL
For Use With (Application) Gene Expression
Hot Start Built-In Hot Start
No. of Reactions 1000 Reactions
Sample Type DNA (Genomic), cDNA
Volume 2 x 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.