missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit (20 mL sample prep, 10 mL PCR)

Product Code. 10045264 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10045264

Brand: Applied Biosystems™ 4403083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Reagents sufficient for 4,000 sample preparation reactions at 50μL/ reaction and 4,000 PCR reactions at 5μL for each reaction.

Go from Biological Sample to Genotyping Results in less than one hour.

The TaqMan™ Sample-to-SNP™ Kit combines DNA extraction and TaqMan™ chemistry-based genotyping in a streamlined protocol to provide results from any sample in less than one hour.

  • DNA Extract All Reagents kits provide PCR-ready DNA from a wide variety of sample types ranging from blood to buccal swabs to plant tissues in 5 min.
  • The TaqMan™ GTXpress™ Master Mix provides robust PCR amplification of extracted DNA in less than 50 min.
  • Compatible with both Fast and Standard mode thermal cycling conditions
  • Pre-mixed components stored at 2° to 8°C significantly reduce assay setup time
  • Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
  • Extended PCR cycling for poorly amplified or sample-limited reactions

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains Lysis Reagents (2° to 8°C): 20 mL each Lysis and DNA Stabilization Solution; Master Mix (15° to 25°C): 10 mL 2X master mix with AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking dye and ROX™ Dye.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
GC-Rich PCR Performance Low
PCR Method qPCR
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast or Standard
Technique SNP Genotyping (Real-Time PCR-Based)
For Use With (Equipment) 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, GeneAmp 9700, StepOne™, Standard Mode, Veriti Thermal Cycler, StepOne™ Fast Mode, StepOne™ Standard Mode
Genotyping Target SNPs (Unknown or Numerous), SNPs (Known)
Passive Reference Dye ROX (Pre-mixed)
Product Line Sample-to-SNP, TaqMan
Product Type Genotyping Reagents
Quantity 1 kit
Sufficient For 400 Reactions
Hot Start Built-In Hot Start
Sample Type Buccal Sample, Cell Cultures, Hair with Follicle, Leaf Punch or Needle, Seed Chip, Tail Biopsy (Mouse), Tail Biopsy (Rat), Tissue (FFPE), Whole Blood
Volume 20 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.