missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Sample-to-SNP™ Kit (20mL Sample prep, 50mL PCR)

Product Code. 10773637 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773637 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10773637 Supplier Applied Biosystems™ Supplier No. 4403087
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Reagents sufficient for 400 sample preparation reactions at 50μL/ reaction and 20,000 PCR reactions at 5μL for each reaction.

Go from Biological Sample to Genotyping Results in less than one hour.

The TaqMan™ Sample-to-SNP™ Kit combines DNA extraction and TaqMan™ chemistry-based genotyping in a streamlined protocol to provide results from any sample in less than one hour.

  • DNA Extract All Reagents kits provide PCR-ready DNA from a wide variety of sample types ranging from blood to buccal swabs to plant tissues in 5 min.
  • The TaqMan™ GTXpress™ Master Mix provides robust PCR amplification of extracted DNA in less than 50 min.
  • Compatible with both Fast and Standard mode thermal cycling conditions
  • Pre-mixed components stored at 2° to 8°C significantly reduce assay setup time
  • Validated with all types of Applied Biosystems™ TaqMan™ Genotyping Assays
  • Extended PCR cycling for poorly amplified or sample-limited reactions

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains Lysis Reagents (2° to 8°C): 20mL each Lysis and DNA Stabilization Solution; Master Mix (15° to 25°C): 50mL 2X master mix with AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking dye and ROX™ Dye.
Detection Method Primer-probe
Polymerase AmpliTaq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, GeneAmp™ 9700, StepOne™, Fast Mode, StepOne™, Standard Mode, Veriti™ Thermal Cycler
Passive Reference Dye ROX (Pre-mixed)
Quantity 1 Kit
Target SNPs (Known), SNPs (Unknown or Numerous)
For Use With (Application) Genotyping
Sample Type Buccal Sample, Cell Cultures, Hair with Follicle, Leaf Punch or Needle, Seed Chip, Tail Biopsy (Mouse), Tail Biopsy (Rat), Tissue (FFPE), Whole Blood

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.