missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ViiA™ 7 Array Card RNaseP Verification Kit

Used to validate the performance of the ViiA™ 7 Real-Time PCR System with the TaqMan™ Low Density Array Block Upgrade.

Brand:  Applied Biosystems™ 4432265

Additional Details : Weight : 0.00450kg

Product Code. 15916065

  • 3100.00 € / 1 set
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: An empty TaqMan Low Density Array and eight vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene. Each vial contains premixed reagents (1X TaqMan Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.

Description

Description

The ViiA™ 7 System TaqMan™ Low Density Array Instrument Verification RNase P kit contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each vial contains premixed reagents (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template. The ViiA™ 7 System TaqMan™ Low Density Array Instrument Verification RNase P kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.

The kit must demonstrate the ability to distinguish between 5000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, the instrument should be verified every six months.

Order Info

Store kit at -15° to -25°C.;

Specifications

Specifications

384-well Microfluidic Card
ViiA™ 7 System
Array Card RNaseP Verification Kit
RNase P Calibration, Instrument Verification Plate
8 x Tubes with Standard Dilutions, Empty Array Card, Port 1 NTC, Port 2 Unknown A, Port 3 U, Store at -15°C to -25°C
ViiA™
Real Time PCR (qPCR)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.