missing translation for 'onlineSavingsMsg'
Learn More

Apramycin sulfate, MedChemExpress

Product Code. 16418736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16418736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418736 Supplier Medchem Express Supplier No. HYB1329
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apramycin sulfate is an aminoglycoside antibiotic mproduced by a strain of Streptomyces tenebrarius, used in veterinary practice.

Apramycin sulfate is an aminoglycoside antibiotic mproduced by a strain of Streptomyces tenebrarius, used in veterinary practice.

Specifications

Chemical Name or Material Apramycin sulfate
CAS 65710-07-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H43N5O15S
Quantity 100 mg
Synonym Nebramycin II sulfate
Solubility Information H2O : ≥ 200 mg/mL (313.65 mM)
SMILES O[C@H]1[C@](O[C@H](O[C@@]([C@H](C[C@H]2N)N)([H])[C@@H]([C@H]2O)O)[C@H](N)C3)([H])[C@@]3([H])O[C@H](O[C@@]([C@@H]([C@@H](O)[C@@H]4N)O)([H])O[C@@H]4CO)[C@H]1NC.O=S(O)(O)=O
Molecular Weight (g/mol) 637.66
Formula Weight 637.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.