missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aprepitant

Product Code. 16697022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16697022 250 mg 250mg
16687022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16697022 Supplier Thermo Scientific Chemicals Supplier No. 465312500
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 170729-80-3 | C23H21F7N4O3, C23 H21 F7 N4 O3 | 534.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170729-80-3
Molecular Formula C23 H21 F7 N4 O3

Specifications

Quantity 250 mg
Solubility practically insoluble
Formula Weight 534.43
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
Chemical Name or Material Aprepitant

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.