missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ APY 29

Description
Allosteric modulator of IRE1α. Inhibits IRE1α autophosphorylation (IC50 = 280 nM) and activates IRE1α ribonuclease activity.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 332.36 |
| Physical Form | Solid |
| Color | White |
| Purity | >98% |
| Molecular Weight (g/mol) | 332.371 |
| Chemical Name or Material | APY 29 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?