missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ AQUADUR™ Sensitive Test Strips

Product Code. 16010852 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Green to Red (Color Change/Total Hardness)
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16010852 Green to Red (Color Change/Total Hardness) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16010852 Supplier Macherey-Nagel™ Supplier No. 91243
  • ONLINE EXCLUSIVE PRICE
    26.10 €
    22.75 € / Pack of 50
    Save 3.35 €
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AQUAUDR™ Sensitive water hardness test strips include all the same benefits as the classic AQUADUR test strips, but feature a special, very low measuring range.

  • Water hardness testing
  • Semi-quantitative test stripsl
  • 2 year shelf life

Specifications

Color Green to Red (Color Change/Total Hardness)
Description Total Hardness Semi-Quantitative Test Strips
Format 50 Test Strips/Tube
Graduations 0 to 5.4 to 10.8 to 18.8 ppm CaCO3
Storage Requirements 4°C to 30°C
Type Test Strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.