Description
The Thermo Scientific™ Aquanex™ is designed to truly help make life easier in lab with an intuitive touch screen interface, easy-access consumables, and smart dispensing features. The easy-to-use interface caters to users that need secured access and seamless event logging, system monitoring, and reporting. A new innovative cartridge design provides low TOC, a longer cartridge life, and an easy-to-change design.
Easily access consumables
- The Aquanex user interface provides step-by-step instructions on how to access and change consumables
- UV lamp, RO assembly, and ultrafilter consumables are easily accessed by removing the magnetic front panel
- No screws or complicated tools are required to change consumables
Auto-detection and event logging
- Aquanex system automatically detects and logs when the DI cartridges are replaced
- The system will notify you when consumables are nearing end of life and require replacement
- Consumable installation and replacement dates can be easily viewed in the maintenance menu
Simplified DI cartridge installation
- No water spills or mess during cartridge installation or removal
- Eject the DI cartridge by simply pressing a button on the side of the system
- Guide rails, built-in sensors, and a quick-connect system allow for smooth installation of DI cartridges
Specifications
Specifications
| Feedwater Source | Tap |
| For Use With (Equipment) | Aquanex Water Purification System |
| Includes | A and B Replacement Cartridges |