Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall
GREENER_CHOICE
Product Code. 30058211
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058211 Medium Pack of 50
30058200 Large Pack of 50
30058284 X-Large Pack of 50
30058309 2X-Large Pack of 50
30058157 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30058211 Supplier Chemsplash Supplier No. 2619.300.005
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    149.09 € / Pack of 50
    Save 1.91 €
    1% Off
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
The Aqua Repel Coverall is made from 40 gsm Polypropylene coated fabric. Is water repellent and provides protection against non-hazardous limited liquid splash.
Chemsplash Aqua Repel Coverall
  • 40 GSM Polypropylene Coated Fabric
  • Non Toxic, Waterproof Fabric
  • Hood With Elasticated Band
  • Elasticated Cuffs and Ankles
  • Half Elasticated Waist
  • Zipped Front With Adhesive Zip Flap
  • PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Specifications
Product Type Hooded Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None