missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall

Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Description
Chemsplash Aqua Repel Coverall
- 40 GSM Polypropylene Coated Fabric
- Non Toxic, Waterproof Fabric
- Hood With Elasticated Band
- Elasticated Cuffs and Ankles
- Half Elasticated Waist
- Zipped Front With Adhesive Zip Flap
- PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | Blue |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction