Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall
Product Code. 30058309
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058309 2X-Large Pack of 50
30058211 Medium Pack of 50
30058200 Large Pack of 50
30058284 X-Large Pack of 50
30058157 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30058309 Supplier Chemsplash Supplier No. 2619.300.008
  • ONLINE EXCLUSIVE PRICE
    160.82 €
    149.09 € / Pack of 50
    Save 11.73 €
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Aqua Repel Coverall is made from 40 gsm Polypropylene coated fabric. Is water repellent and provides protection against non-hazardous limited liquid splash.
Chemsplash Aqua Repel Coverall
  • 40 GSM Polypropylene Coated Fabric
  • Non Toxic, Waterproof Fabric
  • Hood With Elasticated Band
  • Elasticated Cuffs and Ankles
  • Half Elasticated Waist
  • Zipped Front With Adhesive Zip Flap
  • PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Specifications
Product Type Hooded Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None