Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall
GREENER_CHOICE
Product Code. p-200105910
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
Product Code. 30058211
Brand: Chemsplash 2619.300.005
Only null left
Add to basket
Add to basket
The Aqua Repel Coverall is made from 40 gsm Polypropylene coated fabric. Is water repellent and provides protection against non-hazardous limited liquid splash.
Chemsplash Aqua Repel Coverall
  • 40 GSM Polypropylene Coated Fabric
  • Non Toxic, Waterproof Fabric
  • Hood With Elasticated Band
  • Elasticated Cuffs and Ankles
  • Half Elasticated Waist
  • Zipped Front With Adhesive Zip Flap
  • PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Specifications
Product Type Hooded Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Ties
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None