missing translation for 'onlineSavingsMsg'
Learn More

Stuart™ Aquatron™ deionizer Housing and Accessories

Product Code. 16150921
Change view
Click to view available options
Type:
Deionizer Housing
Wall Bracket
Features:
Double
Double Capacity
Single
Single Capacity
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Features unitSize
16150921 Deionizer Housing Double Capacity Each
16170921 Deionizer Housing Single Capacity Each
16160921 Wall Bracket Single Each
16140921 Wall Bracket Double Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16150921 Supplier Stuart™ Supplier No. 8024494
Add to basket
Add to basket
This item is not returnable. View return policy

Stuart™ Aquatron™ deionizer Housing and Accessories provide good, quality water at an affordable price

  • Output up to 60 litres/hour
  • Low cost disposable cartridges
  • Colour change indicates resin condition at a glance
  • Free standing or wall mountable
  • Ideal as pre-treatment for Aquatron™ water stills
  • A simple, portable deionizer giving good quality water at an affordable price
  • The disposable ion exchange cartridges slowly change colour from green to blue as they are exhausted giving at a glance indication of the resin condition
  • No need for conductivity meters, batteries or mains electrical power
  • Supplied complete with flexible hose for connection to a tap and a stopcock to control the flow rate
  • Can be linked in series with the Aquatron™ filter either by a flexible hose or the rigid coupling (ALC) available as an accessory
  • Ideal as a pre-treatment for Aquatron™ water stills to prevent scale build up and improve output quality
  • Housing and ion exchange cartridges must be ordered separately

Specifications

For Use With (Application) Ideal as Pre-Treatment for Aquatron™ Water Stills
Includes Colour change indicator depicts resin condition at a glance
For Use With (Equipment) Aquatron Filter
Type Deionizer Housing
Features Double Capacity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.