missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ AR-GLAS™ Stirring Rods

Constructed of AR-GLAS™. Tubes are fused closely at both ends.

Brand:  BRAND™ 135825

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10014321

  • 74.05 € / Pack of 50

Estimated Shipment: 27-04-2023
to see stock.
Add to basket

Description

Description

  • Constructed of AR-GLAS™
  • Tubes are fused closely at both ends.
  • Supplied in packs of 50
Specifications

Specifications

50/Pk.
Glass
300mm
8mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers