missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic Acid, MP Biomedicals™

Product Code. 11413221 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413221 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413221 Supplier MP Biomedicals Supplier No. 0215038491
  • ONLINE EXCLUSIVE PRICE
    868.00 €
    823.20 € / 1g
    Save 44.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Arachidonic acid is an essential fatty acid, and a precursor in the biosynthesis of prostaglandins, thromboxanes, and leukotrienes. It occurs in liver, brain, glandular organs, and depot fats of animals, in small amounts in human depot fats, and is a cons

Arachidonic acid (AA) is an unsaturated ω-6 fatty acid constituent of the phospholipids of cell membranes. Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation. AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism.

AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis. AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs). Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.

Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin). Arachidonic acid plays a key role in cellular regulation and is controlled through multiple interconnected pathways. Changes in arachidonic acid levels correspond to the synthesis of eicosanoids resulting from receptor-stimulated lipid hydrolysis.It activates protein kinase C. Arachidonic acid serves as a precursor to numerous eicosanoids as well as other bioactive molecules.

Specifications

CAS 506-32-1
Melting Point -49°C (literature)
Density 0.922g/mL (Lit.)
Boiling Point 170°C (0.2 mmHg (literature))
Molecular Formula C20H32O2
Refractive Index n20/D 1.4872 (Lit.)
Quantity 1 g
Synonym arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid
InChI Key YZXBAPSDXZZRGB-DOFZRALJSA-N
SMILES CCCCCC=CCC=CCC=CCC=CCCCC(=O)O
IUPAC Name (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
Molecular Weight (g/mol) 304.474
PubChem CID 444899
ChEBI CHEBI:15843
Formula Weight 304.5
Percent Purity >98%
Grade Not Available
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Arachidonic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: cis,cis,cis,cis-5,8,11,14-eicosatetraenoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.