missing translation for 'onlineSavingsMsg'
Learn More

Aramchol, MedChemExpress

Product Code. 16477908
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16477908

Brand: Medchem Express HY19796

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aramchol (C20-FABAC) is a conjugate of cholic acid and arachidic acid that could inhibit stearoyl coenzyme A desaturase 1 (SCD1) activity. Aramchol has potential use in nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH) treatment.

Aramchol (C20-FABAC) is a conjugate of cholic acid and arachidic acid that could inhibit stearoyl coenzyme A desaturase 1 (SCD1) activity. Aramchol has potential use in nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH) treatment.

Specifications

Chemical Name or Material Aramchol
CAS 246529-22-6
Color White
Purity Grade Notes Research
Molecular Formula C44H79NO5
Quantity 10 mg
Synonym C20-FABAC
Solubility Information DMSO : 25 mg/mL (35.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@H](CCC(O)=O)[C@H]1CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@@H](NC(CCCCCCCCCCCCCCCCCCC)=O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C
Molecular Weight (g/mol) 702.1
Formula Weight 702.1
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.