missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ARC 239 dihydrochloride

Product Code. 18779746
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18779746 25 mg 25mg
18728886 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18779746 Supplier R&D Systems™ Supplier No. 0928/25
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α2B antagonist

ARC 239 dihydrochloride is a selective α2B adrenoceptor antagonist (pKD values are 8.8, 6.7 and 6.4 at α2B, α2A, and α2D receptors respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride
CAS 55974-42-0
Assay Percent Range >98%
Quantity 25 mg
Product Line Tocris
Molecular Formula C24H29N3O3.2HCl
Formula Weight Observed MW: 480.43
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.