missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ARC 239 dihydrochloride
GREENER_CHOICE

Product Code. p-7116025
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18779746

Brand: Tocris Bioscience™ 0928/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α2B antagonist

ARC 239 dihydrochloride is a selective α2B adrenoceptor antagonist (pKD values are 8.8, 6.7 and 6.4 at α2B, α2A, and α2D receptors respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl]-4,4-dimethyl-1,3-(2H,4H)-isoquinolindione dihydrochloride
CAS 55974-42-0
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C24H29N3O3.2HCl
Formula Weight Observed MW: 480.43
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.