missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ArciTect™ Annealing Buffer (5X)

Product Code. 17119311
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17119311 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17119311 Supplier STEMCELL Technologies Supplier No. 76020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For guide RNA generation in CRISPR-Cas9 genome editing

  • ArciTect™ Annealing Buffer (5X) is required to anneal ArciTect™ tracrRNA (Catalog #76016/76017/76018) and ArciTect™ crRNA (Catalog #76010/76011/76012) to form a guide RNA (gRNA) duplex
  • This product is a refill component for ArciTect™ tracrRNA Kit

Specifications

Description Cell Type: Pluripotent Stem Cells
For Use With (Application) Stem Cell Biology
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.