Learn More
STEMCELL Technologies ArciTect™ tracrRNA Kit
Product Code. 17199301
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 nmol Kit
20 nmol Kit
5 nmol Kit
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17199301 10 nmol Kit 1 set
17109311 20 nmol Kit 1 set
17189301 5 nmol Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17199301 Supplier STEMCELL Technologies Supplier No. 76017
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Trans-activating crRNA for guide RNA generation in CRISPR-Cas9 genome editing
  • ArciTect™ tracrRNA is a trans-activating CRISPR RNA (crRNA), one of two RNA components required to make a guide RNA (gRNA) for CRISPR-Cas9 genome editing
  • ArciTect™ tracrRNA contains a complementary sequence to the linker sequence in ArciTect™ crRNA, allowing fast and efficient gRNA duplex formation
  • It is recognized by all ArciTect™ Cas9 nucleases and is essential for ribonucleoprotein (RNP) complex formation
  • It contains 2'-O-methyl and phosphorothioate modifications at the first two 5' and 3' terminal residues for optimal stability and editing efficiency
  • The ArciTect™ tracrRNA Kit includes tracrRNA as well as ArciTect™ Annealing Buffer (5X), which is required for forming a gRNA duplex between ArciTect™ tracrRNA and ArciTect™ crRNA
  • ArciTect™ crRNA is available by custom order for user-specified target sequences, in 2 nmol (Catalog #76010), 10 nmol (Catalog #76011), and 20 nmol (Catalog #76012) sizes
  • ArciTect™ Cas9 Nuclease is also available separately (Catalog #76001)
Specifications
Cell Type Pluripotent Stem Cells
Quantity 10 nmol Kit