missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Arctigenin

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Description
Antioxidant, anti-inflammatory, antiproliferative and antiviral agent. Inhibits LPS-induced iNOS expression via inhibition of IκBα phosphorylation and p65 nuclear translocation (IC50 = 10 nM). Inhibits HIV-1 replication in vitro. Also potently inhibits MKK1 (MEK1) (IC50 = 0.5 nM) and provides neuroprotection by binding to kainate receptors.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 372.42 |
| Purity | >98% |
| Molecular Weight (g/mol) | 372.417 |
| Chemical Name or Material | Arctigenin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction