missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arctiin

Product Code. 16434737
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16434737 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16434737 Supplier MedChemExpress Supplier No. HYN0034
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Arctiin(NSC 315527), a plant lignan that can be extracted from the Arctium lappa (burdock) seeds, is a possible environmental endocrine disruptor compounds and have been shown to influence sex hormone metabolism as well as protein synthesis, steroid biosynthesis.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 534.55 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 20362-31-6
Product Type Natural Products
Purity 98.89%
Solubility DMSO : 250 mg/mL (467.68 mM∣Need ultrasonic) ∣H2O : 10 mg/mL (18.71 mM∣ultrasonic and warming and heat to 60°C)
Synonym Arctii∣ NSC 315527∣ Arctigenin-4-glucoside
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.