missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Arformoterol Tartrate
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 6219/50
This item is not returnable.
View return policy
Description
Arformoterol tartrate is a long-acting β2 adrenergic agonist (LABA). Bronchodilator.Specifications
| Long-acting Agonist (LABA) | |
| 200815-49-2 | |
| 494.49g/mol | |
| Adrenergic beta-2 Receptor Agonists |
| Adrenergic beta-2 | |
| C19H24N2O4.C4H6O6 | |
| Store at 4°C | |
| 50 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only