missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Arg-Gly-Asp

Product Code. 15456909
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456909 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15456909 Supplier Thermo Scientific Chemicals Supplier No. J65468.#0
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cellular and molecular biology applications

Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name RGD
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Fragment from cell-attachment domain of fibronectin; a cell-attachment peptide
Molecular Weight (g/mol) 346.34
Product Type Arg-Gly-Asp
Quantity 5 mg
Regulatory Status RUO - research use only
Sequence Arginyl-glycyl-aspartic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.