missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Argipressin

Product Code. 16453966
Encompass_Preferred
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 16453966

Brand: MedChemExpress HYP0049

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Argipressin (Arg8-vasopressin) binds to the V1, V2, V3-vascular arginine vasopressin receptor, with a Kd value of 1.31 nM in A7r5 rat aortic smooth muscle cells for V1.

Specifications

Purity 99.53%
CAS 113-79-1
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1084.23 g/mol
Product Type Peptides
Quantity 25 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 100 mg/mL (92.23 mM)
Synonym Arg8-vasopressin∣ AVP
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.