missing translation for 'onlineSavingsMsg'
Learn More

Aristolochic Acid A, TRC

Product Code. 30331041
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30331041 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331041 Supplier Toronto Research Chemicals Supplier No. TRCA77130025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aristolochic Acid
CAS 313-67-7
Purity Grade Notes HPLC
Molecular Formula C17 H11 N O7
Quantity 25 mg
Synonym 8-Methoxy-6-nitro-phenanthro[3,4-d]-1,3-dioxole-5-carboxylic acid, 8-Methoxy-3,4-methylenedioxy-10-nitrophenanthrene-1-carboxylic acid, Aristolochia A, Aristolochic acid, Aristolochic acid 1, Aristolochic acid A, Aristolochic acid I, NSC 11926, NSC 50413, TR 1736, Tardolyt
InChI Formula InChI=1S/C17H11NO7/c1-23-12-4-2-3-8-9(12)5-11(18(21)22)14-10(17(19)20)6-13-16(15(8)14)25-7-24-13/h2-6H,7H2,1H3,(H,19,20)
SMILES COc1cccc2c1cc(c3c(cc4OCOc4c23)C(=O)O)[N+](=O)[O-]
IUPAC Name 8-methoxy-6-nitronaphtho[2,1-g][1,3]benzodioxole-5-carboxylic acid
Molecular Weight (g/mol) 341.272
Formula Weight 341.054
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.