missing translation for 'onlineSavingsMsg'
Learn More

Sartorius arium™ mini Scientific Pack

Product Code. 15575153 Shop All Sartorius Products
Change view
Click to view available options
Capacity:
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15575153 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15575153 Supplier Sartorius Supplier No. H2OSPACK
  • ONLINE EXCLUSIVE PRICE
    325.64 €
    302.00 € / Pack of 1
    Save 23.64 €
    7% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with arium mini and arium mini plus Ultrapure Water Systems

  • Deionization cartridge featuring top-down-flow technology
  • High performance capacity thanks to efficient ion exchange resins
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • Best ion removal delivers ultrapure water that exceeds the ASTM Type 1 standard
TRUSTED_SUSTAINABILITY

Specifications

Description Sartorius arium™ mini Ultrapure Water System Accessory, Scientific Kit
Capacity 1 L
Feedwater Source Pretreated
Material Polypropylene
Purification Method Ion Exchange, Activated Carbon
Resin Type Ion Exchange Resin, Activated Carbon
Filter Type Spherical, catalytic activated carbon Ultrapure mixed bed ion exchange resin in semiconductor quality
For Use With (Equipment) arium mini and arium mini plus Ultrapure Water Systems
Type Scientific Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.