missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Smart Station PW
GREENER_CHOICE

Product Code. 17254233 Shop All Sartorius Products
Change view
Click to view available options
Type:
Remote Dispenser Support
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
17254233 Remote Dispenser Support 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17254233 Supplier Sartorius Supplier No. H2OARSTPB
Add to basket
Add to basket
This item is not returnable. View return policy

Arium™ Smart Station flexible remote dispenser for pure water

  • Height adjustable display/dispense station for dispensing pure water
  • Compact design for easy and space-saving integration into lab environment
  • Intuitive touch-activated color display with direct access to important functions
  • Flexible and stepless height adjustment to fill different sized containers and beakers
  • Ergonomic design can be optimized for either right or left hand operation
  • Accurate volume dispense for fast and precise buffer and sample prep
  • Adjustable, splash-free flow rate from drop-by-drop to 2 L/min

Equipment supplied:

  • Display/dispense unit with wall bracket
  • Power supply including:
    • IEC connector
    • Connection set

For intended use with:

  • Arium™ Bagtank
  • Arium™ Pro
  • Arium™ Comfort
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 L
Feed Water Monitoring No
Feedwater Source Pure
Flow Rate Up to 2 L/min.
Product Water Type Pure Water
TOC Monitor with UV Intensity Monitoring No
Type Remote Dispenser Support
Display Touch display
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.