missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Arium™ Smart Station UP
GREENER_CHOICE

Product Code. 17284233 Shop All Sartorius Products
Click to view available options
Type:
Remote Dispenser
Remote Dispenser Support
Unit Size:
1 unit
This item is not returnable. View return policy

Product Code. 17284233

Brand: Sartorius H2OARSTUPT

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Arium™ Smart Station flexible remote dispenser for ultrapure water

  • Height adjustable display/dispense station for dispensing Type 1 ultrapure water
  • Compact design for easy and space-saving integration into lab environment
  • Intuitive touch-activated color display with direct access to important functions
  • Flexible and stepless height adjustment to fill different sized containers and beakers
  • Ergonomic design can be optimized for right or left hand operation
  • Accurate volume dispense for fast and precise buffer and sample prep
  • Adjustable, splash-free flow rate from drop-by-drop to 2 L/min

Equipment supplied:

  • Display/dispense unit with stand
  • Power supply including:
    • IEC connector
    • Connection set

For intended use with:

  • Arium™ Bagtank
  • Arium™ Pro
  • Arium™ Comfort
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 L
Feed Water Monitoring No
Feedwater Source Ultrapure
Flow Rate Up to 2 L/min.
Product Water Type Ultrapure (Type 1)
TOC Monitor with UV Intensity Monitoring No
Type Remote Dispenser
Display Touch display
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.